Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRN/FCGRT Antibody (FCGRT/2932), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB382659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382659 Supplier Novus Biologicals Supplier No. NBP308224AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen FCRN/FCGRT
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone FCGRT/2932
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Host Species Mouse
Immunogen Recombinant fragment of human FCRN/FCGRT protein (around aa 24-215) (Exact sequence is proprietary) (Uniprot: P55899)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.