Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcRn (FCGRT) Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $393.63
Specifications
Antigen | FcRn (FCGRT) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABF191-25UL
|
MilliporeSigma
ABF19125UL |
25 μL |
Each for $192.35
|
|
|||||
ABF191MI
|
MilliporeSigma
ABF191 |
200 μL |
Each for $393.63
|
|
|||||
Description
This rabbit polyclonal Anti-FcRn (FCGRT) antibody detects IgG receptor FcRn large subunit p51. It has been tested for use in Immunofluorescence and Western Blotting.Specifications
FcRn (FCGRT) | |
Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Inflammation and Immunology | |
Fcgrt,Fcrn | |
IgG | |
Affinity Purified | |
This polyclonal antibody detects murine IgG receptor FcRn. It targets an epitope within 44 amino acids from the C-terminal region. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
GST-tagged recombinant fragment corresponding to 44 amino acids from the C-terminal region of murine IgG receptor FcRn large subunit p51. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title