Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Yellow No. 6, Spectrum™ Chemical
SDP

Catalog No. 18602644 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
20 kg
Packaging:
Poly Pail

FD155, 2783-94-0, C16H10N2O7S2Na2

Spectrum™ Chemical FD and C Yellow No. 6

Chemical Identifiers

CAS 2783-94-0
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.36
InChI Key TXVRKNUZLYFDTJ-DDVLFWKVSA-L
IUPAC Name disodium (5E)-6-oxo-5-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-5,6-dihydronaphthalene-2-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(N\N=C2\C(=O)C=CC3=CC(=CC=C23)S([O-])(=O)=O)C=C1

Specifications

CAS Max % 100%
Percent Purity 87%
Grade Reagent
Packaging Poly Pail
Color Orange to Yellow
Quantity 20 kg
Chemical Name or Material FD and C Yellow No. 6
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.