Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fd/M13 bacteriophage Antibody (MM05), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB328678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB328678 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB328678 Supplier Novus Biologicals Supplier No. NBP305788APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

fd/M13 bacteriophage Monoclonal antibody specifically detects fd/M13 bacteriophage in Virus samples. It is validated for ELISA

Specifications

Antigen fd/M13 bacteriophage
Applications ELISA
Classification Monoclonal
Clone MM05
Conjugate APC
Dilution ELISA
Formulation PBS
Gene Alias major coat protein
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with M13 Bacteriophage.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 927333
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.