Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FDFT1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15485520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15485520 20 μL
NBP154855 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15485520 Supplier Novus Biologicals Supplier No. NBP15485520UL

Rabbit Polyclonal Antibody

FDFT1 Polyclonal specifically detects FDFT1 in Human, Bovine samples. It is validated for Western Blot.

Specifications

Antigen FDFT1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 2.5 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P37268
Gene Alias DGPT, EC 2.5.1.21, ERG9, Farnesyl-diphosphate farnesyltransferase, farnesyl-diphosphate farnesyltransferase 1, FPP:FPP farnesyltransferase, presqualene-di-diphosphate synthase, SQS, squalene synthase, squalene synthetase, SS
Gene Symbols FDFT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human FDFT1 (NP_004453). Peptide sequence HSFLYQPDWRFMESKEKDRQVLEDFPTISLEFRNLAEKYQTVIADICRRM.
Molecular Weight of Antigen 48 kDa
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2222
Target Species Human, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.