Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FDPS Antibody (005), FITC, Novus Biologicals™
SDP

Catalog No. NB038063 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038063 Supplier Novus Biologicals Supplier No. NBP290198F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

FDPS Monoclonal specifically detects FDPS in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen FDPS
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 5
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias 6E)-farnesyl diphosphate synthase, Dimethylallyltranstransferase, EC 2.5.1.1, EC 2.5.1.10, farnesyl diphosphate synthaseFPP synthetase, farnesyl pyrophosphate synthase, farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, FPP synthase, FPPS, FPSgeranyltranstransferase, Geranyltranstransferase, KIAA1293
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human FDPS (NP_001129294.1; Met1-Lys353).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2224
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.