Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDPS Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $471.00
Specifications
Antigen | FDPS |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551147
|
Invitrogen™
MA551147 |
200 μL |
Each for $471.00
|
|
|||||
FEMA551148
|
Invitrogen™
MA551148 |
20 μL |
Each for $195.80
|
|
|||||
Description
The isoprene biosynthetic pathway supply the cell with cholesterol, ubiquinone, and various nonsterol metabolites. The farnesylpyrophosphate synthetase enzyme catalyzes the formation of geranyl and farnesylpyrophosphate from isopentenylpyrophosphate and dimethylallyl pyrophosphate. Analysis of FDPS activity and protein in rat liver, accompanied by immunofluorescence and immunoelectron microscopy studies, demonstrated that FDPS is predominantly localized in peroxisomes. 1 Liver tissue from patients with the peroxisomal deficiency diseases Zellweger syndrome and neonatal adrenoleukodystrophy exhibit diminished activities of FDPS and subsequent isoprenoid synthesis.Specifications
FDPS | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
(2E,6E)-farnesyl diphosphate synthase; 6030492I17Rik; Ac2-125; AI256750; cholesterol-regulated 39 kDa protein; CR 39; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyl diphosphate synthase (farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase); farnesyl diphosphate synthetase; farnesyl pyrophosphate synthase; farnesyl pyrophosphate synthetase; farnesyl pyrophosphate synthetase, dimethylallyltranstransferase, geranyltranstransferase; Fdps; Fdpsl1; FPP synthase; FPP synthetase; FPPS; FPS; geranyltranstransferase; KIAA1293; mKIAA1293; POROK9; testis-specific farnesyl pyrophosphate synthetase | |
FDPS | |
IgG1 κ | |
Protein A/G | |
FDPS |
C1 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Rat | |
P05369 | |
83791 | |
Recombinant Rat FDPS protein, Met1-Lys353 (Accession #P05369). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title