Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FDXR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 155841AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-0329


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Ferredoxin reductase is a ubiquitous flavoenzyme, containing noncovalently bound FAD as a prosthetic group (1). It plays a role in delivering NADPH or low potential one-electron donors such as ferredoxin and flavodoxin to redox-based metabolisms in plastids, mitochondria and bacteria (2). In mammals, ferredoxin reductase is loosely associated with the inner mitochondrial membrane and receives electrons from NADPH. These electrons are transferred to ferredoxin which shuttles electrons to cytochrome P450 in the adrenal cortex mitochondrial steroid hydroxylation systems (3).
Specifications

Specifications

FDXR
Polyclonal
Unconjugated
FDXR
Adrenodoxin reductase, ADXR, Aerredoxin Reductase, AR, FDXR, Ferredoxin NADP(+) reductase
Rabbit
Antigen Affinity Chromatography
RUO
14149, 2232, 79122
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22570, P56522, Q61578
Fdxr
FDXR Fusion Protein Ag7145
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.