Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDXR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | FDXR |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0329
![]() ![]() |
Proteintech Group Inc
155841AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Ferredoxin reductase is a ubiquitous flavoenzyme, containing noncovalently bound FAD as a prosthetic group (1). It plays a role in delivering NADPH or low potential one-electron donors such as ferredoxin and flavodoxin to redox-based metabolisms in plastids, mitochondria and bacteria (2). In mammals, ferredoxin reductase is loosely associated with the inner mitochondrial membrane and receives electrons from NADPH. These electrons are transferred to ferredoxin which shuttles electrons to cytochrome P450 in the adrenal cortex mitochondrial steroid hydroxylation systems (3).Specifications
FDXR | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P22570, P56522, Q61578 | |
14149, 2232, 79122 | |
FDXR Fusion Protein Ag7145 | |
Primary | |
-20°C | |
FDXR |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Adrenodoxin reductase, ADXR, Aerredoxin Reductase, AR, FDXR, Ferredoxin NADP(+) reductase | |
Fdxr | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title