Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FE65 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB11058360X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11058360X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11058360X Supplier Novus Biologicals Supplier No. NB11058360AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FE65 Polyclonal specifically detects FE65 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen FE65
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein
Gene Symbols APBB1
Host Species Rabbit
Immunogen Recombinant rat FE65, expressed in bacteria. [Swiss-Prot# P46933]
Molecular Weight of Antigen 100 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 322
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.