Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Herpes Virus Type 1 Antibody (FHV7-7C), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10063151G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063151G 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10063151G Supplier Novus Biologicals Supplier No. NB10063151G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Herpes Virus Type 1 Monoclonal specifically detects Feline Herpes Virus Type 1 in Virus samples. It is validated for ELISA, Western Blot.

Specifications

Antigen Feline Herpes Virus Type 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone FHV7-7C
Conjugate DyLight 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Made to Viral FELINE HERPES VIRUS TYPE 1
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63151 is specific for feline Herpesvirus type 1 (FHV-1), of the genus Herpesviridae. FHV-1 is worldwide, and the most clinically significant pathogen for feline respiratory infections, being the causative agent of feline viral rhinotracheitis (FVR). Clone FHV7-7C specifically reacts with three different cultured FHV isolates, but does not cross react with feline Calcivirus (FCV), feline Immunodeficiency virus (FIV) or feline Leukaemia virus (FeLV).
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.