Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Herpes Virus Type 1 Antibody (FHV7-7C), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB209212 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB209212 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB209212 Supplier Novus Biologicals Supplier No. NB10063151DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Herpes Virus Type 1 Monoclonal antibody specifically detects Feline Herpes Virus Type 1 in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Feline Herpes Virus Type 1
Applications Western Blot, ELISA
Classification Monoclonal
Clone FHV7-7C
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Made to Viral FELINE HERPES VIRUS TYPE 1
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.