Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Immunodeficiency Virus Antibody (NM3-6A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100665000.1MG
Description
Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Feline Immunodeficiency Virus | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:25-1:200, Flow Cytometry 1:50-1:100, ELISA 0.5 ug/well | |
CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24 | |
Native, extracts | |
0.1 mg | |
Primary | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA | |
NM3-6A9 | |
Unconjugated | |
PBS (pH 7.2) with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Recognizes Feline Immunodeficieny Virus (FIV) p24 gag derived from p50 GAG polyprotein. This does not recognize Feline Leukemia Virus, Feline Herpes Virus type 1, Feline Coronavirus or Feline Calicivirus. | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction