Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Immunodeficiency Virus Antibody (NM3-6A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Feline Immunodeficiency Virus |
---|---|
Clone | NM3-6A9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:25-1:200, Flow Cytometry 1:50-1:100, ELISA 0.5 ug/well |
Applications | Western Blot, ELISA, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017778
![]() |
Novus Biologicals
NB100665000.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB100665001
![]() |
Novus Biologicals
NB100665000.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
Feline Immunodeficiency Virus | |
1 mg/mL | |
Western Blot, ELISA, Flow Cytometry | |
Unconjugated | |
Mouse | |
Virus | |
CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24 | |
IgG1 | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
NM3-6A9 | |
Western Blot 1:25-1:200, Flow Cytometry 1:50-1:100, ELISA 0.5 ug/well | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.2) with 0.05% Sodium Azide | |
Native, extracts | |
Primary | |
Protein A purified | |
Recognizes Feline Immunodeficieny Virus (FIV) p24 gag derived from p50 GAG polyprotein. This does not recognize Feline Leukemia Virus, Feline Herpes Virus type 1, Feline Coronavirus or Feline Calicivirus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title