Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Immunodeficiency Virus Antibody (NM3-6A9), DyLight 350, Novus Biologicals™
SDP

Catalog No. N10650UV01M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N10650UV01M 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N10650UV01M Supplier Novus Biologicals Supplier No. NB10066500UV0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Immunodeficiency Virus Monoclonal specifically detects Feline Immunodeficiency Virus in Virus samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen Feline Immunodeficiency Virus
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone NM3-6A9
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CA; Capsid protein p24; Feline immunodeficiency virus; Feline immunodeficiency virus capsid protein p24; Feline immunodeficiency virus p24; FIV capsid protein p24
Host Species Mouse
Immunogen Native, extracts
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognizes Feline Immunodeficieny Virus (FIV) p24 gag derived from p50 GAG polyprotein. This does not recognize Feline Leukemia Virus, Feline Herpes Virus type 1, Feline Coronavirus or Feline Calicivirus.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.