Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Immunodeficiency Virus p24 gag Antibody (PAK3-2C1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB10064802J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064802J 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064802J Supplier Novus Biologicals Supplier No. NB10064802APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Immunodeficiency Virus p24 gag Monoclonal specifically detects Feline Immunodeficiency Virus p24 gag in Virus samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Feline Immunodeficiency Virus p24 gag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone PAK3-2C1
Conjugate APC
Dilution Flow Cytometry
Gene Alias CA, Capsid protein p24, Feline immunodeficiency virus, Feline immunodeficiency virus capsid protein p24, Feline immunodeficiency virus p24, FIV capsid protein p24
Host Species Mouse
Immunogen Made to Viral FELINE IMMUNODEFICIENCY VIRUS p24 gag
Purification Method Affinity Purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64802 recognizes an immunodominant p24 epitope of feline immunodeficiency virus (FIV) p24 gag. The lentivirus FIV, responsible for a progressive and debilitating immune deficiency syndrome in domestic cats, similar to that caused by the human Immunodeficiency Virus (HIV), is a complex retrovirus with a tightly-packed genome, containing the structural genes gag, env and pro-pol and the accessory genes vif, rev and ORF-A/2. The specific binding epitope recognized by clone PAK3-2C1 appears to be enclosed in a tight peptide coil during gag production and maturation. The addition of 0.3% Triton-X detergent is recommended to reveal this epitope for antibody binding. NB100-64802 detects a dominant band of approximately 24kDa, and also detects p24 gag precursor bands at 36, 39, 49 and 52kDa under reducing conditions in Western blotting. NB100-64802 does not recognize Feline Leukaemia Virus (FeLV), Feline Herpes Virus (FHV) type 1, Feline Coronavirus or Feline Calicivirus.
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.