Learn More
Feline Immunodeficiency Virus p24 gag Antibody (PAK3-2C1), DyLight 550, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Feline Immunodeficiency Virus p24 gag |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | PAK3-2C1 |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | CA, Capsid protein p24, Feline immunodeficiency virus, Feline immunodeficiency virus capsid protein p24, Feline immunodeficiency virus p24, FIV capsid protein p24 |
| Host Species | Mouse |
| Immunogen | Made to Viral FELINE IMMUNODEFICIENCY VIRUS p24 gag |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.