Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Immunodeficiency Virus p24 gag Antibody (PAK3-2C1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB212578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212578 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212578 Supplier Novus Biologicals Supplier No. NB10064802JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Immunodeficiency Virus p24 gag Monoclonal antibody specifically detects Feline Immunodeficiency Virus p24 gag in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen Feline Immunodeficiency Virus p24 gag
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone PAK3-2C1
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CA, Capsid protein p24, Feline immunodeficiency virus, Feline immunodeficiency virus capsid protein p24, Feline immunodeficiency virus p24, FIV capsid protein p24
Host Species Mouse
Immunogen Made to Viral FELINE IMMUNODEFICIENCY VIRUS p24 gag
Purification Method Protein A or G purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.