Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukaemia Virus gp70 Antibody (C11D8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB211105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211105 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211105 Supplier Novus Biologicals Supplier No. NB10065565AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Leukaemia Virus gp70 Monoclonal antibody specifically detects Feline Leukaemia Virus gp70 in Virus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Feline Leukaemia Virus gp70
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone C11D8
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Env polyprotein, Feline Leukemia Virus glycoprotein 70, Glycoprotein gp70 [Precursor], GP70 envelope protein, gp70 protein (SU), p15E protein, p15E protein (TM)
Host Species Mouse
Immunogen Made to Viral FELINE LEUKAEMIA VIRUS gp70
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.