Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukaemia Virus gp70 Antibody (C11D8), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB10065565C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065565C 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065565C Supplier Novus Biologicals Supplier No. NB10065565C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Leukaemia Virus gp70 Monoclonal specifically detects Feline Leukaemia Virus gp70 in Virus samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Feline Leukaemia Virus gp70
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C11D8
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Env polyprotein, Feline Leukemia Virus glycoprotein 70, Glycoprotein gp70 [Precursor], GP70 envelope protein, gp70 protein (SU), p15E protein, p15E protein (TM)
Host Species Mouse
Immunogen Made to Viral FELINE LEUKAEMIA VIRUS gp70
Quantity 0.125 mL
Primary or Secondary Primary
Test Specificity NB100-65565 recognizes the gp85/gp70 envelope glycoprotein of feline leukaemia virus. The antibody specifically recognizes the epitope defined by amino acids 214-218 of the gp70 sequence (aa MGPNL). Some weak cross-reactivity with the sequence MGPLL found in the FeLV p15e membrane protein may also occur. This antibody effectively neutralises all three FeLV subgroups (A,B and C).
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.