Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukaemia Virus p15E Antibody (PF6J-2A1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB10065808K Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065808K 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065808K Supplier Novus Biologicals Supplier No. NB10065808FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Leukaemia Virus p15E Monoclonal specifically detects Feline Leukaemia Virus p15E in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Feline Leukaemia Virus p15E
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone PF6J-2A1
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Env polyprotein, Feline Leukemia Virus glycoprotein 70, Glycoprotein gp70 [Precursor], GP70 envelope protein, gp70 protein (SU), p15E protein, p15E protein (TM)
Host Species Mouse
Immunogen Made to Viral FELINE LEUKAEMIA VIRUS p15E
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65808 recognizes an epitope within the amino-terminal domain of both the 20kDa full-length (including intracellular sequence), and the 15kDa truncated forms, of feline leukaemia virus (FeLV) p15e transmembrane envelope protein. Persistent infection of cats with the oncoretrovirus FeLV can result in the non-neoplastic condition known as FeLV-associated enteritis (FAE). Studies have shown strong expression of the major FeLV proteins p15e and gp70 in intestinal crypt epithelial cells of cats with FAE, compared to FeLV-positive cats without intestinal alterations, thereby suggesting a pathogenic role for these envelope proteins/precursors in FAE.
Reconstitution Protein A purified
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.