Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukaemia Virus p27 Antibody (PF12J-10A), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB211148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211148 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211148 Supplier Novus Biologicals Supplier No. NB10065688AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Leukaemia Virus p27 Monoclonal antibody specifically detects Feline Leukaemia Virus p27 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Feline Leukaemia Virus p27
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PF12J-10A
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias feline leukemia provirus, Feline leukemia virus p27, Feline leukemia virus subgroup c receptor related protein 1, Feline sarcoma virus, flvcr protein, flvcr1, FPS, hflvcr, Oncogene fes
Host Species Mouse
Immunogen Made to Viral FELINE LEUKAEMIA VIRUS p27
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.