Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline Leukaemia Virus p27 Antibody (PF12J-10A), FITC, Novus Biologicals™
SDP

Catalog No. NB10065688F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065688F 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065688F Supplier Novus Biologicals Supplier No. NB10065688F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Feline Leukaemia Virus p27 Monoclonal specifically detects Feline Leukaemia Virus p27 in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Feline Leukaemia Virus p27
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone PF12J-10A
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias feline leukemia provirus, Feline leukemia virus p27, Feline leukemia virus subgroup c receptor related protein 1, Feline sarcoma virus, flvcr protein, flvcr1, FPS, hflvcr, Oncogene fes
Host Species Mouse
Immunogen Made to Viral FELINE LEUKAEMIA VIRUS p27
Purification Method Protein A purified
Quantity 0.125 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65688 recognizes feline leukaemia virus (FeLV) p27 core protein (p27). The oncoretrovirus FeLV is responsible for both neoplastic and non-neoplastic conditions, often resulting in a poor prognosis and fatal outcome for cats persistently infected. During persistent infection the virus, soluble viral antigens and antigen containing white blood cells are continuously present in the blood and diagnosis is mainly based upon detection of the p27 antigen - using ELISA to test for p27 as it circulates free in the bloodstream, and Immunofluorescence studies to detect p27 within the infected white cells.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.