Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FEN-1 Antibody (OTI1F3), Biotin, Novus Biologicals™
SDP

Catalog No. NBP270716B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270716B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270716B Supplier Novus Biologicals Supplier No. NBP270716B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FEN-1 Monoclonal specifically detects FEN-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FEN-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1F3
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.1, FEN-1hFEN-1, flap structure-specific endonuclease 1DNase IV, Maturation factor 1, maturation factor-1, MF1flap endonuclease 1, RAD2
Gene Symbols FEN1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 117-380 of human FEN-1 (NP_004102) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Base Excision Repair, Cancer, DNA Repair, Editing and Processing Endonucleases
Primary or Secondary Primary
Gene ID (Entrez) 2237.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.