Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fenoldopam hydrochloride
GREENER_CHOICESDP

Catalog No. 165910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-591-0 10 mg
16-595-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 16-591-0 Supplier Tocris Bioscience™ Supplier No. 1659/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective D1-like partial agonist

Selective D1-like dopamine receptor partial agonist (EC50 = 57 nM). Vasodilator in vivo and does not readily cross the blood-brain barrier. Also α2-adrenoceptor antagonist in vitro (Ki = 15 - 25 nM).

Chemical Identifiers

CAS 181217-39-0
Molecular Formula C16H17Cl2NO3

Specifications

Quantity 10 mg
Formula Weight 342.22
Purity >98%
Molecular Weight (g/mol) 342.216
Chemical Name or Material Fenoldopam hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.