Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Fenoldopam hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
Selective D1-like dopamine receptor partial agonist (EC50 = 57 nM). Vasodilator in vivo and does not readily cross the blood-brain barrier. Also α2-adrenoceptor antagonist in vitro (Ki = 15 - 25 nM).
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 342.22 |
Purity | >98% |
Molecular Weight (g/mol) | 342.216 |
Chemical Name or Material | Fenoldopam hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction