Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fer Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP120089Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP120089Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP120089Y Supplier Novus Biologicals Supplier No. NBP120089AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fer Polyclonal specifically detects Fer in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen Fer
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P16591
Gene Alias EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer
Gene Symbols FER
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot# P16591]
Molecular Weight of Antigen 94 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2241
Test Specificity Human. Predicted to react with mouse and canine based on 100% sequence homology.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.