Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fer Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB338497 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338497 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338497 Supplier Novus Biologicals Supplier No. NBP120089MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Fer Polyclonal antibody specifically detects Fer in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, KnockDown

Specifications

Antigen Fer
Applications Western Blot, Immunocytochemistry, KnockDown
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, EC 2.7.10.2, fer (fps/fes related) tyrosine kinase, p94-Fer, Proto-oncogene c-Fer, TYK3phosphoprotein NCP94, Tyrosine kinase 3, tyrosine-protein kinase Fer
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human FER (within residues 400-450). [Swiss-Prot# P16591]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2241
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.