Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FER1L5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120403
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5120403 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5120403

Supplier: Invitrogen™ PA5120403

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Rat testis. The target is usually found in the following locations: Cell membrane, Membrane, Single-pass membrane protein. Immunogen sequence: KLPTLSEKKH QDFLGYLYRK FWFKSSKAED EYEHEVDWWS KLFWATDEHK SLKYKYKDYH TLKVYECELE AVPAFQGLQD FCQTFKLYQE QPKLDSPVVG EFKGLFRIYP FPENPEAPKP PLQFLVWPER EDFPQPCLVR VYMVRAINLQ PQDYNGLCDP YVILKLGKTE LGNRDMYQPN TLDPIFGMMF ELTCNIPLEK DLEIQLYDFD LFSPDDKIGT TVIDLENRLL SGFGAHCGLS KSYCQSGPFR WRDQMPPSYL LERYAKRKGL PPPLFSPEED AVFYNGKKFK LQSFEPKTPT VHGLGPKKER LALYLLHTQG L.

Plays a role in myoblast fusion; probable mediator of endocytic recycling for membrane trafficking events during myotube formation. [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FER1L5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Fer1l5
Gene Accession No. P0DM40
Gene Symbols Fer1l5
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100534273, 679806
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.