Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bsp143I (Sau3AI) (10 U/μL)

Catalog No. p-4521536
Click to view available options
Quantity:
1,500 units
300 units

Cut at ^GATC sites with Bsp143I (Sau3AI), which performs best at 37°C in its own unique buffer (Isoschizomers: BfuCI, BssMI, BstKTI, BstMBI, DpnII, Kzo9I, NdeII, Sau3AIm).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...▵G A T C ...3'

3'... C T A G▵...5'

Conditions for 100% Activity:

  • 1X Buffer Bsp143I: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Bsp143I is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Bsp143I more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: completely overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — blocked
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — no effect

Note:

DpnI, Bsp143I and MboI all recognize the same sequence but have different methylation sensitivities and cleavage sites. Compatible Ends: BamHI, BclI, BglII, PsuI.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Bsp143I (Sau3AI)
Compatible Buffer Unique Buffer (10X Buffer Bsp143I (Sau3AI))
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 300 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.