Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Cfr9I (XmaI) (10 U/μL)
GSA_VA

Catalog No. p-4521671
Click to view available options
Quantity:
1,500 units
300 units

The Cfr9I (XmaI) restriction enzyme recognizes C^CCGGG sites and cuts best at 37°C in its own unique buffer (Isoschizomers: TspMI, XmaCI, XmaI).

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C▵C C G G G...3'

3'...G G G C C▵C...5'

Conditions for 100% Activity:

  • 1X Buffer Cfr9I:10mM Tris-HCl (pH 7.2 at 37°C), 5mM MgCl2, 200mM sodium glutamate and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Cfr9I is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 250mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Cfr9I, more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: completely overlaps-cleavage impaired
  • EcoKI: never overlaps-no effect
  • EcoBI: never overlaps-no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

To achieve complete digestion of substrate with Cfr9I, the concentration of DNA should be no less than 50μg/mL in the reaction buffer.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Cfr9I (XmaI)
Compatible Buffer Unique Buffer (10X Buffer Cfr9I (XmaI))
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 300 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.