Learn More
Thermo Scientific™ EpiJET DNA Methylation Analysis Kit (MspI/HpaII)
Description
5-Methylcytosine is a prominent epigenetic DNA modification which plays an important role in regulation of gene expression.
EpiJET DNA Methylation Analysis Kit (MspI/HpaII) uses the MspI and HpaII restriction enzymes to analyze DNA methylation status at a specific locus. Epi MspI and Epi HpaII are isoschizomers with differing sensitivities to CpG methylation. When the internal CpG in the 5'-CCGG-3' tetranucleotide sequence is methylated, cleavage with Epi HpaII is blocked, but cleavage with Epi MspI is not affected.
The Epi MspI and Epi HpaII enzymes are specially formulated for epigenetics studies to complete genomic DNA digestion in 1 hour.
Highlights:
• Fast—complete digestion of genomic DNA in 1 hour at 37°C
• Efficient—specially formulated enzymes for epigenetic studies of DNA methylation status
Specifications
Specifications
| Compatible Buffer | 10X Epi Buffer |
| Quantity | 200 reactions |
| Product Type | DNA Methylation Analysis Kit |
| Product Line | EpiJET™ |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.