Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ GsuI (BpmI) (5 U/μL)

Catalog No. p-4521809
Click to view available options
Quantity:
100 units
500 units

The GsuI (BpmI) restriction enzyme recognizes CTGGAG(16/14)^ sites and cuts best at 30°C in B buffer (Isoschizomers: BpmI).

Lambda DNA digested with GsuI (BpmI), 1% agarose, 25 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...C T G G A G (N)16▵...3'

3'...G A C C T C (N)14▵...5'

Conditions for 100% Activity:

  • 1X Buffer B: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 and 0.1mg/mL BSA
  • Incubate at 30°C

Storage Buffer:

  • GsuI is supplied in:10mM potassium phosphate (pH 7.5 at 25°C), 1mM EDTA, 1mM DTT, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: may overlap —blocked
  • CpG: never overlaps —no effect
  • EcoKI: never overlaps —no effect
  • EcoBI: never overlaps —no effect

Note:

  • Incubation at 37°C results in 70% activity
  • GsuI requires only Mg2 + for its activity, but is stimulated by S-adenosylmethionine
  • 0.01mM S-adenosylmethionine gives a 2-fold increase in activity
  • GsuI may remain associated with the cleaved DNA
  • This may cause DNA band shifting during electrophoresis
  • To avoid atypical DNA band patterns, use the 6X DNA Loading Dye and SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis
  • GsuI is blocked by overlappingdcm methylation
  • To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099)
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 U/μL
Methylation Sensitivity Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme GsuI (BpmI)
Compatible Buffer 10X Buffer B
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 30°C
Type IIS RE Yes
Quantity 100 units
Product Type Restriction Enzyme
Research Category Scarless Cloning
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.