Learn More
Thermo Scientific™ HinfI (10 U/μL)
Description
Lambda DNA digested with HinfI, 1.4% agarose, 148 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...G▵A N T C...3'
3'...C T N A▵G...5' Conditions for 100% Activity:
- 1X Buffer R: 10mM Tris-HCl (pH8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- HinfI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: may overlap —cleavage impaired
- EcoKI: never overlaps —no effect
- EcoBI: may overlap —blocked
Compatible Ends:
- G▴AWTC — PfeI

Specifications
Specifications
Concentration | 10 U/μL |
Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
Enzyme | Hinf I |
Compatible Buffer | 10X Buffer R |
Sensitive to Heat Inactivation | Yes |
Optimal Reaction Temperature | 37°C |
Type IIS RE | No |
Quantity | 2,000 units |
Product Type | Restriction Enzyme |
Research Category | Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.