Learn More
Thermo Scientific™ HpaII (10 U/μL)
Description
Lambda DNA digested with HpaII, 1.4% agarose, 328 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...C▵C G G...3'
3'...G G C▵C...5' Conditions for 100% Activity:
- 1X Buffer Tango™: 33mM Tris-acetate (pH7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- HpaII is supplied in: 10mM Tris-HCl (pH7.4 at 25°C), 100mM NaCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: never overlaps —no effect
- CpG: completely overlaps —blocked
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Compatible Ends:
- Bsp119I, Bsu15I, Hin1I, MaeII, NarI, Psp1406I, SsiI, TaqI, XmiI

Specifications
Specifications
Concentration | 10 U/μL |
Methylation Sensitivity | CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive |
Enzyme | Hpa II |
Compatible Buffer | 10X Buffer Tango |
Sensitive to Heat Inactivation | Yes |
Optimal Reaction Temperature | 37°C |
Type IIS RE | No |
Quantity | 5,000 units |
Product Type | Restriction Enzyme |
Research Category | Traditional Cloning |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.