Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial
GSA_VA

Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.

Supplier:  Thermo Scientific™ K0252

 View more versions of this product

Catalog No. FERK0252


Only null left
Edge
Add to Cart

Description

Description

The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.

  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X
SYBR
High
Taq DNA Polymerase
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Real Time PCR SYBR Master Mix
1000 Reactions at 25 μL
1,000 Reactions
10 x 1.25 mL
Contains:
• 10 x 1.25 mL Maxima SYBR Green qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 1000 x 25 μL reactions.
• 250 μL ROX solution, 50 μM
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Tube
qPCR
Standard
Maxima
1,000 reactions
Gene Expression
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only. Not for use in diagnostic procedures.