Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MluI (10 U/μL)

Catalog No. p-4521836
Change view
Click to view available options
Quantity:
1,000 units
5,000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERER0561 1,000 units
FERER0562 5,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERER0561 Supplier Thermo Scientific™ Supplier No. ER0561
Only null left
Add to Cart
Edge
Add to Cart

The MluI restriction enzyme recognizes A^CGCGT sites and cuts best at 37°C in R buffer.

Lambda DNA digested with MluI, 0.7% agarose, 7 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A▵C G C G T...3'

3'...T G C G C▵A...5'

Conditions for 100% Activity:

  • 1X Buffer R: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • MluI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: may overlap —no effect
  • EcoBI: may overlap —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Compatible Ends:

  • AflIII, BtgI, PauI, SgsI
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme Mlu I
Compatible Buffer 10X Buffer R
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE No
Quantity 1,000 units
Product Type Restriction Enzyme
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.