Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RevertAid First Strand cDNA Synthesis Kit
GSA_VA

Catalog No. p-4526935
Click to view available options
No. of Reactions:
100 Reactions
20 Reactions

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates.

Thermo Scientific RevertAid First Strand cDNA Synthesis Kit is a complete system for efficient synthesis of first strand cDNA from RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), a recombinant M-MuLV RT which maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA templates from degradation at temperatures up to 55°C.

The RevertAid First Strand cDNA Synthesis Kit is supplied with both oligo(dT)18 and random hexamer primers. The oligo(dT)18 primer anneals selectively to the poly(A) tail of mRNA. Random hexamer primers do not require the presence of the poly(A) tail; they can therefore be used for transcription of the 5'-end regions of mRNA or cDNA synthesis of RNA species lacking a poly(A) tail (e.g., microRNAs). Gene-specific primers may also be used with the kit.

Radioactively- and non-radioactively-labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid First Strand cDNA Synthesis Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit with all components necessary for the RT reaction already included

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

• RevertAid Reverse Transcriptase
• RiboLock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Oligo(dT)18 Primer
• Random Hexamer Primer
• Control GAPDH RNA
• Forward GAPDH Primer (10 μM)
• Reverse GAPDH Primer (10 μM)
• Nuclease-free water

Store at –20°C.

Format Kit
Reaction Speed 60 min.
Technique Reverse Transcription
Optimal Reaction Temperature 42°C
Quantity 20 reactions
Reverse Transcriptase RevertAid
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) RT-PCR
Final Product Type First-Strand cDNA
No. of Reactions 20 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 13 kb
Starting Material RNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.