Learn More
American Educational Products Fermentation Tubes
SCI_ED
Catalog No. s33839
Click to view available options
Quantity:
12/Pk.
Trap gas generated by the fermentation process.
These Durham tubes consist of 150 × 16mm tubes with 75 × 10mm tubes to be inverted inside the larger tubes to trap gas generated by the fermentation process. Sold in packs of 12.
Specifications
Material Glass
Quantity 12/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.