Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FERMT3/URP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | FERMT3/URP2 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
FERMT3/URP2 Polyclonal specifically detects FERMT3/URP2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
FERMT3/URP2 | |
Polyclonal | |
Rabbit | |
Human | |
fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3 | |
FERMT3 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
83706 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MADSSYTSEVQAILAFLSLQRTGSGGPGNHPHGPDASAEGLNPYGLVAPRFQRKFKAKQLT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title