Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FERMT3/URP2 Antibody (OTI3C7), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP272241A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272241A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272241A30 Supplier Novus Biologicals Supplier No. NBP272241AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FERMT3/URP2 Monoclonal specifically detects FERMT3/URP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FERMT3/URP2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3C7
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias fermitin family member 3, KIND3fermitin family homolog 3 (Drosophila), kindlin 3, Kindlin-3, MGC10966, MIG-2, MIG2BURP2SF, MIG2-like protein, UNC-112 related protein 2, UNC112C, Unc-112-related protein 2, URP2fermitin family homolog 3
Gene Symbols FERMT3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 259-528 of human FERMT3/URP2 (NP_113659) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 83706
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.