Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Ammonium Citrate, Brown, Powder, FCC, 16.5-18.5%, Spectrum™ Chemical
SDP

Catalog No. 18602656 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail

Catalog No. 18-602-656

Supplier: Spectrum Chemical Manufacturing Corporation F1000125GM

F1000, 1185-57-5

Spectrum™ Chemical Ferric Ammonium Citrate, Brown, Powder, FCC is a brownish powder which is easily soluble in water. It is a food additive used as an acidity regulator and anti-caking agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H8FeNO7
Molecular Weight (g/mol) 261.98
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-K
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
SMILES N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 16.5 to 18.5%
Grade FCC
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.