Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferric Citrate, Purified, Spectrum™ Chemical
SDP

Catalog No. 18602667 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-602-667

Supplier: Spectrum Chemical Manufacturing Corporation F1028125GM

Only null left
Add to Cart
Add to Cart

F1028, 2338-05-8, Approx. FeC6H5O7·xH2O

Spectrum™ Chemical Ferric Citrate, Purified is a powder with a green to reddish-brown appearance. It can also be used in water purification, printing and an acidity regulator.

Chemical Identifiers

CAS 2338-05-8
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
IUPAC Name iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 125 g
Grade Purified
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.