Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Antibody (F23), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB1108384AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1108384AF 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1108384AF Supplier Novus Biologicals Supplier No. NB1108384AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for ELISA, Sandwich ELISA.

Specifications

Antigen Ferritin
Applications ELISA
Classification Monoclonal
Clone F23
Conjugate Alexa Fluor 405
Dilution ELISA, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Gene Symbols FTH1
Host Species Mouse
Immunogen This clone has been derived from hybridization of SP2/0 myeloma cells with spleen cells of Balb/c mice immunised with purified human ferritin.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2495
Test Specificity Ferritin (F23)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.