Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Antibody (FTH/2081), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB391924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391924 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391924 Supplier Novus Biologicals Supplier No. NBP308581UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ferritin Monoclonal antibody specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Ferritin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FTH/2081
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Host Species Mouse
Immunogen Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary) (Uniprot: P02794)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2495
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.