Learn More
Ferritin Heavy Chain Antibody (962610), Alexa Fluor™ 594, R&D Systems™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Ferritin Heavy Chain |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 962610 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, Immunocytochemistry/Immunofluorescence, Immunocytochemistry |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 |
| Gene Symbols | FTH1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.