Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Light Chain Antibody (FTL/1388) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254584100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254584100 100 μg
NB254584200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254584100 Supplier Novus Biologicals Supplier No. NBP254584100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ferritin Light Chain Monoclonal specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.

Specifications

Antigen Ferritin Light Chain
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone FTL/1388
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.1 - 0.2 ug/million cells, Immunocytochemistry/Immunofluorescence 0.1 - 0.2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3
Gene Symbols FTL
Host Species Mouse
Immunogen Recombinant human FTL protein fragment (aa 38-165) (exact sequence is proprietary)
Molecular Weight of Antigen 22 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2512
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.