Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferritin Light Chain Antibody (FTL/2338R), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB391756 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391756 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391756 Supplier Novus Biologicals Supplier No. NBP308620AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ferritin Light Chain Monoclonal antibody specifically detects Ferritin Light Chain in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Ferritin Light Chain
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone FTL/2338R
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias Ferritin L subunit, ferritin L-chain, ferritin light chain, ferritin light polypeptide-like 3, ferritin, light polypeptide, MGC71996, NBIA3
Host Species Rabbit
Immunogen Recombinant human Ferritin Light Chain protein fragment (around aa 38-165) (exact sequence is proprietary) (Uniprot: P02792)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 2512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.