Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroportin/SLC40A1 Antibody (1308C), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N275923APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N275923APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N275923APC7 Supplier Novus Biologicals Supplier No. NBP275923APCCY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Ferroportin/SLC40A1 Monoclonal specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Ferroportin/SLC40A1
Applications Flow Cytometry
Classification Monoclonal
Clone 1308C
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1
Gene Symbols SLC40A1
Host Species Rabbit
Immunogen This Ferroportin/SLC40A1 Antibody (1308C) [Allophycocyanin/Cy7] was developed against a synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-320). [UniProt# Q9NP59]
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30061.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.