Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ferroportin/SLC40A1 Antibody (8G10NB), FITC, Novus Biologicals™
SDP

Catalog No. NB271254 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271254 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271254 Supplier Novus Biologicals Supplier No. NBP245357F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ferroportin/SLC40A1 Monoclonal antibody specifically detects Ferroportin/SLC40A1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Intracellular)

Specifications

Antigen Ferroportin/SLC40A1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8G10NB
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular)
Formulation PBS
Gene Alias Ferroportin-1, FPN1, FPN1IREG1ferroportin 1, HFE4, HFE4ferroportin-1, IREG1, iron regulated gene 1, Iron-regulated transporter 1, member 3, MST079, MSTP079, MTP1, putative ferroportin 1 variant IIIB, SLC11A3, SLC11A3iron regulated gene 1, solute carrier family 11 (proton-coupled divalent metal ion transporters), solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3, solute carrier family 40 (iron-regulated transporter), member 1, solute carrier family 40 member 1
Host Species Mouse
Immunogen This Ferroportin/SLC40A1 Antibody (8G10NB) was developed against partial recombinant human Ferroportin protein (between amino acids 150-350) [Uniprot: Q9NP59].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 30061
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.